Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other JAK2 SIRNA Assays ›
          Gene Symbol:
          JAK2
          Gene Name:
          Janus kinase 2
          Gene Aliases:
          JTK10, THCYT3
          Chromosome Location:
          Chr.9: 4985086 - 5128183 on Build GRCh38
          UniGene ID:
          Hs.656213
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 145133

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: JAK2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001322194.1 NP_001309123.1 21 3442
          NM_001322195.1 NP_001309124.1 20 2960
          NM_001322196.1 NP_001309125.1 20 3273
          NM_001322198.1 NP_001309127.1 21 3261
          NM_001322199.1 NP_001309128.1 21 3203
          NM_001322204.1 NP_001309133.1 18 2712
          NM_004972.3 NP_004963.1 21 3341
          GenBank mRNA AF001362.1 - 19 2847
          AF005216.1 - 21 3341
          AF058925.1 - 21 3324
          AK292525.1 - 20 2960
          AK302618.1 - 18 2771
          AY973034.1 - 20 2918
          BC039695.1 - 20 2945 5556026

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          MAPK cascade
          activation of MAPKK activity
          adaptive immune response
          protein phosphorylation
          apoptotic process
          activation of cysteine-type endopeptidase activity involved in apoptotic process
          movement of cell or subcellular component
          signal transduction
          enzyme linked receptor protein signaling pathway
          G-protein coupled receptor signaling pathway
          positive regulation of cytosolic calcium ion concentration
          JAK-STAT cascade
          tyrosine phosphorylation of STAT protein
          STAT protein import into nucleus
          mesoderm development
          blood coagulation
          positive regulation of cell proliferation
          negative regulation of cell proliferation
          intrinsic apoptotic signaling pathway in response to oxidative stress
          negative regulation of cardiac muscle cell apoptotic process
          positive regulation of cell-substrate adhesion
          positive regulation of phosphatidylinositol 3-kinase signaling
          cell migration
          peptidyl-tyrosine phosphorylation
          cytokine-mediated signaling pathway
          negative regulation of cell-cell adhesion
          actin filament polymerization
          cell differentiation
          erythrocyte differentiation
          positive regulation of cell migration
          axon regeneration
          mineralocorticoid receptor signaling pathway
          positive regulation of insulin secretion
          response to lipopolysaccharide
          positive regulation of phosphoprotein phosphatase activity
          positive regulation of interleukin-1 beta production
          positive regulation of tumor necrosis factor production
          positive regulation of protein import into nucleus, translocation
          response to hydroperoxide
          tumor necrosis factor-mediated signaling pathway
          response to tumor necrosis factor
          histone H3-Y41 phosphorylation
          intracellular signal transduction
          interleukin-12-mediated signaling pathway
          peptidyl-tyrosine autophosphorylation
          regulation of cell proliferation
          tyrosine phosphorylation of Stat3 protein
          tyrosine phosphorylation of Stat5 protein
          tyrosine phosphorylation of Stat1 protein
          positive regulation of tyrosine phosphorylation of Stat3 protein
          positive regulation of tyrosine phosphorylation of Stat5 protein
          activation of JAK2 kinase activity
          regulation of apoptotic process
          positive regulation of DNA binding
          negative regulation of DNA binding
          negative regulation of neuron apoptotic process
          positive regulation of GTPase activity
          innate immune response
          positive regulation of nitric oxide biosynthetic process
          positive regulation of cell differentiation
          negative regulation of heart contraction
          response to antibiotic
          protein autophosphorylation
          platelet-derived growth factor receptor signaling pathway
          regulation of inflammatory response
          positive regulation of inflammatory response
          positive regulation of peptidyl-tyrosine phosphorylation
          positive regulation of cell activation
          positive regulation of sequence-specific DNA binding transcription factor activity
          positive regulation of nitric-oxide synthase biosynthetic process
          interferon-gamma-mediated signaling pathway
          regulation of interferon-gamma-mediated signaling pathway
          growth hormone receptor signaling pathway
          JAK-STAT cascade involved in growth hormone signaling pathway
          positive regulation of growth hormone receptor signaling pathway
          mammary gland epithelium development
          response to interleukin-12
          extrinsic apoptotic signaling pathway
          activation of cysteine-type endopeptidase activity involved in apoptotic signaling pathway
          protein kinase activity
          protein tyrosine kinase activity
          non-membrane spanning protein tyrosine kinase activity
          Ras guanyl-nucleotide exchange factor activity
          receptor binding
          growth hormone receptor binding
          interleukin-12 receptor binding
          protein binding
          ATP binding
          protein C-terminus binding
          protein kinase binding
          heme binding
          type 1 angiotensin receptor binding
          acetylcholine receptor binding
          histone kinase activity (H3-Y41 specific)
          SH2 domain binding
          histone binding
          phosphatidylinositol 3-kinase binding
          insulin receptor substrate binding
          metal ion binding
          peptide hormone receptor binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-mtqfw:80/100.66.79.68:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0