Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ELAVL2 SIRNA Assays ›
          Gene Symbol:
          ELAVL2
          Gene Name:
          ELAV like neuron-specific RNA binding protein 2
          Gene Aliases:
          HEL-N1, HELN1, HUB
          Chromosome Location:
          Chr.9: 23690099 - 23850757 on Build GRCh38
          UniGene ID:
          Hs.166109
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 146481

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: ELAVL2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001171195.1 NP_001164666.1 4, 5 744
          NM_001171197.1 NP_001164668.1 4, 5 757
          NM_004432.3 NP_004423.2 4, 5 763
          XM_005251393.3 XP_005251450.1 4, 5 901
          XM_005251394.3 XP_005251451.1 4, 5 1058
          XM_005251395.3 XP_005251452.1 4, 5 838
          XM_006716734.3 XP_006716797.1 5, 6 1150
          XM_006716735.3 XP_006716798.1 5, 6 618
          XM_006716736.3 XP_006716799.1 4, 5 1039
          XM_011517774.2 XP_011516076.1 5, 6 1164
          XM_011517776.2 XP_011516078.1 4, 5 594
          XM_011517777.2 XP_011516079.1 5, 6 1150
          XM_011517778.2 XP_011516080.2 4, 5 687
          XM_011517779.2 XP_011516081.1 5, 6 662
          XM_011517780.2 XP_011516082.1 5, 6 991
          XM_011517783.2 XP_011516085.1 5, 6 1024
          XM_011517784.2 XP_011516086.1 4, 5 719
          XM_011517785.2 XP_011516087.1 4, 5 524
          XM_011517786.2 XP_011516088.1 4, 5 898
          XM_017014408.1 XP_016869897.1 6, 7 1165
          XM_017014409.1 XP_016869898.1 5, 6 803
          XM_017014410.1 XP_016869899.1 5, 6 947
          XM_017014411.1 XP_016869900.1 5, 6 1130
          XM_017014412.1 XP_016869901.1 6, 7 1112
          XM_017014413.1 XP_016869902.1 6, 7 1111
          XM_017014414.1 XP_016869903.1 5, 6 1134
          XM_017014415.1 XP_016869904.1 4, 5 580
          XM_017014416.1 XP_016869905.1 5, 6 653
          XM_017014417.1 XP_016869906.1 5, 6 1024
          XM_017014418.1 XP_016869907.1 4, 5 858
          XM_017014419.1 XP_016869908.1 4, 5 749
          XM_017014420.1 XP_016869909.1 4, 5 752
          XM_017014421.1 XP_016869910.1 4, 5 870
          XM_017014422.1 XP_016869911.1 4, 5 1039
          XM_017014423.1 XP_016869912.1 4, 5 696
          XM_017014424.1 XP_016869913.1 4, 5 1042
          XM_017014425.1 XP_016869914.1 4, 5 850
          XM_017014426.1 XP_016869915.1 3, 4 379
          GenBank mRNA AB209294.1 - 4, 5 763
          BC030692.1 - 4, 5 761 4826082
          BC042393.1 - 4, 5 747

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: XM_006238385, XM_006238388, XM_008763850, XM_008763848, XM_006238389, and others.
          This siRNA also targets the following transcripts in Mouse: NM_001177883, NM_207686, NM_207685, NM_010486, U29088, and others.

          Panther Classification:

          Molecular Function -

          translation initiation factor

          Gene Ontology Categories:

          Function(s) Process(es)

          mRNA splicing, via spliceosome
          regulation of transcription, DNA-templated
          nucleotide binding
          mRNA 3'-UTR binding
          protein binding
          poly(A) RNA binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-qlhjl:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0