Search
Search

Nsmaf| Gene Symbol: Nsmaf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010945.2 | NP_035075.2 | 3 | 396 | ||
| GenBank mRNA | AF013632.1 | - | 3 | 389 | ||
| AK039902.1 | - | 3 | 247 | |||
| AK049887.1 | - | 3 | 381 | |||
| AK134518.1 | - | 3 | 151 | |||
| AK147709.1 | - | 3 | 256 | |||
| AK164305.1 | - | 3 | 396 | |||
| BC066075.1 | - | 3 | 378 | |||
| BC082810.1 | - | 3 | 378 | |||
| BC145961.1 | - | 3 | 210 | 40131264 | ||
| BC145963.1 | - | 3 | 210 | 40131266 | ||
| BY729997.1 | - | 3 | 397 | |||