Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atcay | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178662.3 | NP_848777.1 | 4 | 513 | ||
GenBank mRNA | AK028298.1 | - | 4 | 520 | ||
AK038483.1 | - | 4 | 523 | |||
AK038990.1 | - | 4 | 514 | |||
AK039411.1 | - | 4 | 521 | |||
AK086723.1 | - | 4 | 555 | |||
AK162959.1 | - | 4 | 521 | |||
AY349150.1 | - | 4 | 478 | |||
BC048903.1 | - | 4 | 497 | 6491141 |