Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atcay | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178662.3 | NP_848777.1 | 13 | 1677 | ||
GenBank mRNA | AK028298.1 | - | 13 | 1684 | ||
AK038483.1 | - | 13 | 1687 | |||
AK038990.1 | - | 13 | 1678 | |||
AK039411.1 | - | 13 | 1685 | |||
AK086723.1 | - | 13 | 1720 | |||
AK162959.1 | - | 13 | 1685 | |||
BC048903.1 | - | 13 | 1661 | 6491141 |