Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other APOBEC2 SIRNA Assays ›
Gene Symbol:
Apobec2
Gene Name:
apolipoprotein B mRNA editing enzyme, catalytic polypeptide 2
Gene Aliases:
Arp1
Chromosome Location:
Chr.17: 48419231 - 48432728 on Build GRCm38
UniGene ID:
Mm.281793
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID
160724
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Apobec2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_009694.3
NP_033824.1
2
754
GenBank mRNA
AF161699.1
-
2
755
AK009047.1
-
2
652
BC027530.1
-
2
664
1433092
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_001106883, FQ215792, FQ216808, BC168177, FQ214919, and others.
Panther Classification:
Molecular Function -
RNA processing factor
Gene Ontology Categories:
Function(s)
Process(es)
mRNA processing
mRNA modification
DNA demethylation
catalytic activity
GTP cyclohydrolase activity
tRNA-specific adenosine deaminase activity
zinc ion binding
hydrolase activity
hydrolase activity, acting on carbon-nitrogen (but not peptide) bonds, in cyclic amidines
N-cyclopropylmelamine deaminase activity
N-cyclopropylammeline deaminase activity
N-cyclopropylammelide alkylamino hydrolase activity
2,5-diamino-6-ribitylamino-4(3H)-pyrimidinone 5'-phosphate deaminase activity
tRNA-specific adenosine-37 deaminase activity
archaeal-specific GTP cyclohydrolase activity
metal ion binding
tRNA-specific adenosine-34 deaminase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-zjghb:80/100.66.79.197:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0