Search
Search

Cpt1a| Gene Symbol: Cpt1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013495.2 | NP_038523.2 | 4 | 391 | ||
| GenBank mRNA | AF017175.1 | - | 279 | |||
| AK050213.1 | - | 4 | 684 | |||
| AK077454.1 | - | 4 | 345 | |||
| AK128925.1 | - | 4 | 345 | |||
| AK133472.1 | - | 4 | 346 | |||
| AK136487.1 | - | 4 | 345 | |||
| AK149688.1 | - | 4 | 342 | |||
| BC038395.1 | - | 3 | 326 | 2649352 | ||
| BC046383.1 | - | 3 | 326 | 4160676 | ||
| BC054791.1 | - | 4 | 372 | 6832731 | ||
| D49472.1 | - | 3 | 318 | |||