Search
Search

GUCA1C| Gene Symbol: GUCA1C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005459.3 | NP_005450.3 | 1 | 218 | ||
| XM_005247910.3 | XP_005247967.1 | 1 | 294 | |||
| GenBank mRNA | AB593147.1 | - | 1 | 165 | ||
| AF110002.1 | - | 1 | 161 | |||
| AF110003.1 | - | 1 | 89 | |||
| AV732101.1 | - | 1 | 218 | |||
| BC103993.1 | - | 1 | 161 | 40022396 | ||
| BC103994.1 | - | 1 | 161 | 40022398 | ||