Search
Search

Zdhhc15| Gene Symbol: Zdhhc15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175358.4 | NP_780567.1 | 10 | 1166 | ||
| GenBank mRNA | AK029137.1 | - | 10 | 906 | ||
| AK032922.1 | - | 10 | 887 | |||
| AK077949.1 | - | 10 | 915 | |||
| AK132304.1 | - | 10 | 1168 | |||
| AK160360.1 | - | 10 | 906 | |||