Search
Search

Rhog| Gene Symbol: Rhog | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019566.3 | NP_062512.1 | 2 | 299 | ||
| GenBank mRNA | AB025943.1 | - | 1 | 157 | ||
| AK143623.1 | - | 2 | 292 | |||
| AK149848.1 | - | 2 | 292 | |||
| AK169853.1 | - | 2 | 283 | |||
| BC059775.1 | - | 2 | 277 | 5370916 | ||
| BQ942269.1 | - | 2 | 299 | |||
| EF524070.1 | - | 2 | 251 | |||