Search
Search

Rhog| Gene Symbol: Rhog | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019566.3 | NP_062512.1 | 2 | 945 | ||
| GenBank mRNA | AK143623.1 | - | 2 | 938 | ||
| AK149848.1 | - | 2 | 938 | |||
| AK169853.1 | - | 2 | 929 | |||
| AK188210.1 | - | 1 | 423 | |||
| BC059775.1 | - | 2 | 924 | 5370916 | ||