Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ndufs8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271443.1 | NP_001258372.1 | 6 | 498 | ||
NM_001271444.1 | NP_001258373.1 | 6 | 738 | |||
NM_144870.5 | NP_659119.2 | 6 | 741 | |||
GenBank mRNA | AK135069.1 | - | 6 | 499 | ||
AK149674.1 | - | 6 | 699 | |||
AK153182.1 | - | 6 | 685 | |||
AK153900.1 | - | 6 | 475 | |||
AK164901.1 | - | 6 | 475 | |||
AK167907.1 | - | 6 | 476 | |||
BC086766.1 | - | 6 | 452 | 6820786 |