Search
Search

Fam13b| Gene Symbol: Fam13b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146084.1 | NP_666196.1 | 3 | 637 | ||
| GenBank mRNA | AK029766.1 | - | 3 | 629 | ||
| AK054416.1 | - | 1 | 1043 | |||
| AK084711.1 | - | 3 | 646 | |||
| BC031465.1 | - | 3 | 637 | 3981146 | ||