Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trerf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001097623.1 | NP_001091092.1 | 19 | 4286 | ||
NM_172622.2 | NP_766210.1 | 16 | 4063 | |||
GenBank mRNA | AK046822.1 | - | 14 | 3704 | ||
AK190506.1 | - | 1 | 96 | |||
AY081774.1 | - | 15 | 4009 | |||
BC051645.1 | - | 5 | 906 | |||
BC059215.1 | - | 19 | 4286 | 6412734 | ||
BC108416.1 | - | 14 | 3746 | 5697521 |