Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other PRMT5 SIRNA Assays ›
          Gene Symbol:
          PRMT5
          Gene Name:
          protein arginine methyltransferase 5
          Gene Aliases:
          HRMT1L5, IBP72, JBP1, SKB1, SKB1Hs
          Chromosome Location:
          Chr.14: 22920511 - 22929585 on Build GRCh38
          UniGene ID:
          Hs.367854
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 17375

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: PRMT5
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001039619.2 NP_001034708.1 2 472
          NM_001282953.1 NP_001269882.1 2 472
          NM_001282955.1 NP_001269884.1 2 387
          NM_001282956.1 NP_001269885.1 2 472
          NM_006109.4 NP_006100.2 2 387
          GenBank mRNA AB451246.1 - 2 163
          AB451370.1 - 2 163
          AF015913.1 - 2 163
          AF167572.1 - 2 254
          AK075251.1 - 2 265
          AK301812.1 - 2 278
          AK302240.1 - 2 167
          AK315500.1 - 2 203
          AL833612.1 - 2 172
          BC025979.2 - 2 175 3833019
          CR456741.1 - 2 163

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_001108867, CB814013.
          This siRNA also targets the following transcripts in Mouse: NM_013768, AF167573, AK076346, AK084106, AK165165, and others.

          Panther Classification:

          Molecular Function -

          protein modifying enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          spliceosomal snRNP assembly
          DNA-templated transcription, termination
          regulation of transcription, DNA-templated
          regulation of mitotic nuclear division
          cell proliferation
          peptidyl-arginine methylation
          peptidyl-arginine methylation, to symmetrical-dimethyl arginine
          circadian regulation of gene expression
          peptidyl-arginine N-methylation
          endothelial cell activation
          histone H4-R3 methylation
          regulation of DNA methylation
          negative regulation of cell differentiation
          positive regulation of oligodendrocyte differentiation
          regulation of ERK1 and ERK2 cascade
          Golgi ribbon formation
          liver regeneration
          regulation of signal transduction by p53 class mediator
          negative regulation of nucleic acid-templated transcription
          positive regulation of adenylate cyclase-inhibiting dopamine receptor signaling pathway
          core promoter sequence-specific DNA binding
          transcription corepressor activity
          protein binding
          methyltransferase activity
          methyl-CpG binding
          histone-arginine N-methyltransferase activity
          protein-arginine N-methyltransferase activity
          protein complex binding
          protein-arginine omega-N symmetric methyltransferase activity
          ribonucleoprotein complex binding
          histone methyltransferase activity (H4-R3 specific)
          protein heterodimerization activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-n469w:80/100.66.75.150:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0