Search
Search

AIMP2| Gene Symbol: AIMP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001326606.1 | NP_001313535.1 | 4 | 739 | ||
| NM_001326607.1 | NP_001313536.1 | 3 | 652 | |||
| NM_001326609.1 | NP_001313538.1 | 5 | 945 | |||
| NM_001326611.1 | NP_001313540.1 | 4 | 658 | |||
| NM_006303.3 | NP_006294.2 | 4 | 859 | |||
| XM_005249847.3 | XP_005249904.1 | 4 | 1073 | |||
| XM_005249848.2 | XP_005249905.1 | 4 | 749 | |||
| XM_005249850.3 | XP_005249907.1 | 4 | 670 | |||
| XM_006715772.2 | XP_006715835.1 | 4 | 644 | |||
| XM_011515519.1 | XP_011513821.1 | 4 | 674 | |||
| GenBank mRNA | AK313037.1 | - | 4 | 865 | ||
| BC002853.1 | - | 4 | 819 | 3634481 | ||
| BC010156.1 | - | 4 | 819 | 3619946 | ||
| BC013630.1 | - | 4 | 827 | 3446074 | ||
| CX867709.1 | - | 4 | 711 | |||
| U24169.1 | - | 4 | 859 | |||