Search
Search

Tti1| Gene Symbol: Tti1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029282.1 | NP_083558.1 | 1 | 748 | ||
| GenBank mRNA | AF332067.1 | - | 1 | 731 | ||
| AF332068.1 | - | 1 | 731 | |||
| AK128940.1 | - | 2 | 798 | |||
| BC053067.1 | - | 1 | 748 | 5698252 | ||