Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpatch1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026181.1 | NP_080457.1 | 13 | 1886 | ||
GenBank mRNA | AK004868.1 | - | 13 | 1886 | ||
AK014367.1 | - | 13 | 1865 | |||
AK154571.1 | - | 13 | 1867 | |||
BC026394.1 | - | 7 | 1052 | |||
BC100577.1 | - | 13 | 1861 | |||
BC125584.1 | - | 13 | 1877 | 40130099 | ||
BC132155.1 | - | 13 | 1877 | 40130432 |