Search
Search

Arfgap3| Gene Symbol: Arfgap3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025445.4 | NP_079721.2 | 9 | 798 | ||
| GenBank mRNA | AK007732.1 | - | 9 | 718 | ||
| AK028990.1 | - | 9 | 799 | |||
| AK075788.1 | - | 9 | 787 | |||
| AK169107.1 | - | 9 | 708 | |||
| AK215301.1 | - | 4 | 175 | |||
| BC004081.1 | - | 9 | 734 | 3592719 | ||
| BC060369.1 | - | 9 | 725 | 6310619 | ||