Search
Search

Extl3| Gene Symbol: Extl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018788.3 | NP_061258.2 | 3 | 1018 | ||
| GenBank mRNA | AF083550.1 | - | 1 | 811 | ||
| AK137842.1 | - | 1 | 95 | |||
| AK220344.1 | - | 1 | 361 | |||
| BC058526.1 | - | 1 | 408 | |||
| BC065073.1 | - | 1 | 483 | 6834580 | ||