Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other WNT4 SIRNA Assays ›
Gene Symbol:
Wnt4
Gene Name:
wingless-type MMTV integration site family, member 4
Gene Aliases:
Wnt-4
Chromosome Location:
Chr.4: 137277635 - 137299501 on Build GRCm38
UniGene ID:
Mm.20355
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID
187049
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
4,126.00
Your Price
Online offer:
4126.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Wnt4
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_009523.2
NP_033549.1
2
178
GenBank mRNA
AK012727.1
-
2
305
M89797.1
-
2
178
Back To Top
More Information
Panther Classification:
Molecular Function -
intercellular signal molecule
Gene Ontology Categories:
Function(s)
Process(es)
metanephros development
branching involved in ureteric bud morphogenesis
mesonephros development
embryonic epithelial tube formation
androgen biosynthetic process
signal transduction
cell-cell signaling
multicellular organism development
gamete generation
sex differentiation
male gonad development
female gonad development
cellular response to starvation
negative regulation of gene expression
negative regulation of steroid biosynthetic process
Wnt signaling pathway
regulation of cell-cell adhesion
cell differentiation
neuron differentiation
female sex determination
negative regulation of cell migration
positive regulation of bone mineralization
positive regulation of aldosterone biosynthetic process
positive regulation of collagen biosynthetic process
T cell differentiation in thymus
immature T cell proliferation in thymus
tube morphogenesis
non-canonical Wnt signaling pathway
non-canonical Wnt signaling pathway via MAPK cascade
negative regulation of fibroblast growth factor receptor signaling pathway
hormone metabolic process
positive regulation of GTPase activity
cell fate commitment
negative regulation of cell differentiation
positive regulation of osteoblast differentiation
positive regulation of meiotic nuclear division
negative regulation of transcription, DNA-templated
positive regulation of transcription, DNA-templated
oocyte development
branching morphogenesis of an epithelial tube
anatomical structure development
smooth muscle cell differentiation
positive regulation of stress fiber assembly
positive regulation of focal adhesion assembly
canonical Wnt signaling pathway
somatotropin secreting cell differentiation
thyroid-stimulating hormone-secreting cell differentiation
mesenchymal to epithelial transition
tertiary branching involved in mammary gland duct morphogenesis
kidney morphogenesis
negative regulation of wound healing
positive regulation of dermatome development
paramesonephric duct development
negative regulation of testicular blood vessel morphogenesis
nephron development
renal vesicle formation
renal vesicle induction
metanephric mesenchymal cell differentiation
mesonephric tubule development
metanephric tubule formation
metanephric nephron development
metanephric nephron morphogenesis
establishment of protein localization to plasma membrane
negative regulation of canonical Wnt signaling pathway
positive regulation of canonical Wnt signaling pathway
negative regulation of male gonad development
positive regulation of cortisol biosynthetic process
negative regulation of testosterone biosynthetic process
negative regulation of apoptotic signaling pathway
transcription corepressor activity
receptor binding
frizzled binding
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0