Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001284308.1 | NP_001271237.1 | 5 | 440 | ||
NM_001284309.1 | NP_001271238.1 | 5 | 380 | |||
NM_001284310.1 | NP_001271239.1 | 4 | 355 | |||
NM_001284311.1 | NP_001271240.1 | 3 | 208 | |||
NM_006869.3 | NP_006860.1 | 5 | 613 | |||
GenBank mRNA | AB097049.1 | - | 5 | 587 | ||
AF082324.1 | - | 5 | 587 | |||
AJ006422.1 | - | 5 | 609 | |||
AK092471.1 | - | 4 | 357 | |||
AK124979.1 | - | 3 | 459 | |||
AK290517.1 | - | 6 | 559 | |||
AK300644.1 | - | 3 | 192 | |||
AK300859.1 | - | 3 | 195 | |||
AK300999.1 | - | 5 | 440 | |||
BC033747.1 | - | 5 | 414 | 5195416 | ||
BM837987.1 | - | 3 | 208 | |||
DA872042.1 | - | 5 | 380 |