Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other XCL1 SIRNA Assays ›
Gene Symbol:
Xcl1
Gene Name:
chemokine (C motif) ligand 1
Gene Aliases:
Ltn, Scyc1
Chromosome Location:
Chr.13: 83199402 - 83202845 on Build Rnor_6.0
UniGene ID:
Rn.90962
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
193440
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Xcl1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_134361.1
NP_599188.1
3
198
GenBank mRNA
U23377.1
-
3
198
Back To Top
More Information
Panther Classification:
Molecular Function -
cytokine
intercellular signal molecule
Gene Ontology Categories:
Function(s)
Process(es)
positive regulation of T cell mediated cytotoxicity
positive regulation of leukocyte chemotaxis
negative regulation of T cell cytokine production
positive regulation of T cell cytokine production
chemotaxis
immune response
cell-cell signaling
response to virus
positive regulation of T cell chemotaxis
negative regulation of interferon-gamma production
negative regulation of interleukin-2 production
positive regulation of interleukin-10 production
mature natural killer cell chemotaxis
negative regulation of sequence-specific DNA binding transcription factor activity
negative regulation of transcription, DNA-templated
regulation of inflammatory response
release of sequestered calcium ion into cytosol
positive regulation of release of sequestered calcium ion into cytosol
cell chemotaxis
cellular response to interleukin-4
cellular response to transforming growth factor beta stimulus
positive regulation of transforming growth factor beta production
positive regulation of granzyme B production
positive regulation of neutrophil chemotaxis
positive regulation of thymocyte migration
positive regulation of natural killer cell chemotaxis
positive regulation of granzyme A production
negative regulation of T-helper 1 cell activation
positive regulation of B cell chemotaxis
positive regulation of T-helper 2 cell cytokine production
positive regulation of T-helper 1 cell cytokine production
positive regulation of immunoglobulin production in mucosal tissue
negative regulation of CD4-positive, alpha-beta T cell proliferation
positive regulation of CD4-positive, alpha-beta T cell proliferation
positive regulation of CD8-positive, alpha-beta T cell proliferation
chemokine activity
chemokine receptor binding
protein homodimerization activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-zjghb:80/100.66.79.197:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0