Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other FLT4 SIRNA Assays ›
          Gene Symbol:
          FLT4
          Gene Name:
          fms related tyrosine kinase 4
          Gene Aliases:
          FLT-4, FLT41, LMPH1A, PCL, VEGFR-3, VEGFR3
          Chromosome Location:
          Chr.5: 180601506 - 180650298 on Build GRCh38
          UniGene ID:
          Hs.646917
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 195

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: FLT4
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_002020.4 NP_002011.2 3 369
          NM_182925.4 NP_891555.2 3 369
          XM_011534478.2 XP_011532780.1 3 747
          XM_017009263.1 XP_016864752.1 3 753
          XM_017009264.1 XP_016864753.1 3 753
          XM_017009265.1 XP_016864754.1 3 754
          XM_017009266.1 XP_016864755.1 3 748
          XM_017009267.1 XP_016864756.1 3 748
          XM_017009268.1 XP_016864757.1 3 312
          GenBank mRNA AK291679.1 - 3 345
          AK309910.1 - 3 345
          AY233382.1 - 3 310
          AY233383.1 - 3 310
          DB455035.2 - 3 369
          EU826564.1 - 3 290
          EU826565.1 - 3 290
          EU826566.1 - 3 290
          U43143.1 - 3 320
          X68203.1 - 3 309
          X69878.1 - 3 311

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of protein phosphorylation
          positive regulation of endothelial cell proliferation
          vasculature development
          lymph vessel development
          lymphangiogenesis
          sprouting angiogenesis
          transmembrane receptor protein tyrosine kinase signaling pathway
          positive regulation of cell proliferation
          positive regulation of vascular endothelial growth factor production
          positive regulation of endothelial cell migration
          peptidyl-tyrosine phosphorylation
          cellular response to vascular endothelial growth factor stimulus
          vascular endothelial growth factor signaling pathway
          negative regulation of apoptotic process
          positive regulation of MAPK cascade
          positive regulation of JNK cascade
          protein autophosphorylation
          vascular endothelial growth factor receptor signaling pathway
          blood vessel morphogenesis
          regulation of blood vessel remodeling
          positive regulation of ERK1 and ERK2 cascade
          positive regulation of protein kinase C signaling
          transmembrane receptor protein tyrosine kinase activity
          vascular endothelial growth factor-activated receptor activity
          protein binding
          ATP binding
          growth factor binding
          protein phosphatase binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-x88k6:80/100.66.75.98:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0