Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other TPT1 SIRNA Assays ›
          Gene Symbol:
          Tpt1
          Gene Name:
          tumor protein, translationally-controlled 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.15: 57891680 - 57894504 on Build Rnor_6.0
          UniGene ID:
          Rn.36610
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          siRNA ID 195798

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Tpt1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_053867.1 NP_446319.1 3 268
          GenBank mRNA BC086358.1 - 3 279 7303675
          FQ210153.1 - 3 296
          FQ210845.1 - 2 182
          FQ217312.1 - 3 296
          FQ217485.1 - 3 296
          FQ220662.1 - 3 296
          FQ220913.1 - 3 299
          FQ221184.1 - 3 296
          FQ221191.1 - 3 285
          FQ221220.1 - 3 296
          FQ221227.1 - 3 296
          FQ221357.1 - 3 296
          FQ221503.1 - 3 296
          FQ221524.1 - 3 296
          FQ221585.1 - 3 296
          FQ221695.1 - 3 296
          FQ221696.1 - 3 297
          FQ222704.1 - 3 295
          FQ222827.1 - 3 296
          FQ223078.1 - 3 296
          FQ223172.1 - 3 297
          FQ223294.1 - 3 303
          FQ223538.1 - 3 296
          FQ223612.1 - 3 285
          FQ224095.1 - 3 296
          FQ224271.1 - 3 297
          FQ224320.1 - 3 295
          FQ224622.1 - 3 232
          FQ224742.1 - 3 296
          FQ226252.1 - 3 296
          FQ227715.1 - 3 297
          FQ227806.1 - 3 325
          FQ228514.1 - 3 296
          FQ228556.1 - 3 296
          FQ228624.1 - 3 296
          FQ228630.1 - 3 296
          FQ228644.1 - 3 293
          FQ229028.1 - 3 296
          FQ229093.1 - 3 296
          FQ229217.1 - 3 296
          FQ229279.1 - 3 296
          FQ229346.1 - 3 296
          FQ229353.1 - 3 296
          FQ230785.1 - 3 296
          FQ231778.1 - 3 296
          FQ232593.1 - 3 296
          FQ232768.1 - 3 297
          FQ233320.1 - 3 327
          FQ233909.1 - 3 299
          FQ234047.1 - 3 286
          FQ234186.1 - 3 297
          FQ234298.1 - 3 296
          FQ234781.1 - 3 296
          FQ234990.1 - 3 296
          FQ235307.1 - 3 296
          U20525.1 - 3 268

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Mouse: NM_009429, AK152690, AK165736, AK152808, AK159565, and others.

          Panther Classification:

          Molecular Function -

          non-motor microtubule binding protein

          Gene Ontology Categories:

          Function(s) Process(es)

          spermatogenesis
          cell proliferation
          response to virus
          stem cell population maintenance
          negative regulation of intrinsic apoptotic signaling pathway in response to DNA damage
          negative regulation of ectoderm development
          calcium ion binding
          transcription factor binding
          poly(A) RNA binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0