Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ASIC2 SIRNA Assays ›
Gene Symbol:
Asic2
Gene Name:
acid-sensing (proton-gated) ion channel 2
Gene Aliases:
Accn1, BNC1k, BNaC1, MDEG, MDEG1, MDEG2
Chromosome Location:
Chr.10: 68247800 - 69347223 on Build Rnor_6.0
UniGene ID:
Rn.37523
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
197233
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Asic2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001034014.1
NP_001029186.1
1529
NM_012892.2
NP_037024.2
1544
GenBank mRNA
U53211.1
-
1529
Y14635.1
-
1544
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Human: NM_001094, NM_183377, U57352, BC075043, U53212, and others.
This siRNA also targets the following transcripts in Mouse: NM_007384, NM_001034013, U57353, AF348465, AK134778, and others.
Panther Classification:
Molecular Function -
ion channel
Gene Ontology Categories:
Function(s)
Process(es)
regulation of systemic arterial blood pressure by aortic arch baroreceptor feedback
cation transport
sodium ion transport
phototransduction
sensory perception of sound
response to mechanical stimulus
response to acidic pH
regulation of gene expression
regulation of vasoconstriction
regulation of blood coagulation
ion transmembrane transport
regulation of ion transmembrane transport
protein localization to synapse
cellular response to drug
sodium ion transmembrane transport
regulation of membrane potential
negative regulation of apoptotic process
sensory perception of sour taste
detection of mechanical stimulus involved in sensory perception
positive regulation of synapse assembly
cellular response to acidic pH
cation transmembrane transport
positive regulation of cation channel activity
ion channel activity
voltage-gated sodium channel activity
cation channel activity
ligand-gated sodium channel activity
ion gated channel activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-vnb2r:80/100.66.79.197:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0