Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ARRB2 SIRNA Assays ›
Gene Symbol:
Arrb2
Gene Name:
arrestin, beta 2
Gene Aliases:
BARRES
Chromosome Location:
Chr.10: 57040252 - 57048045 on Build Rnor_6.0
UniGene ID:
Rn.32973
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
197276
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Arrb2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_012911.1
NP_037043.1
4, 5
352
GenBank mRNA
BC087578.1
-
4, 5
224
7311618
M91590.1
-
4, 5
352
Back To Top
More Information
Panther Classification:
Molecular Function -
scaffold/adaptor protein
Gene Ontology Categories:
Function(s)
Process(es)
regulation of protein phosphorylation
positive regulation of protein phosphorylation
G-protein coupled receptor internalization
desensitization of G-protein coupled receptor protein signaling pathway by arrestin
positive regulation of receptor internalization
transcription from RNA polymerase II promoter
endocytosis
transforming growth factor beta receptor signaling pathway
G-protein coupled receptor signaling pathway
brain development
adult walking behavior
regulation of G-protein coupled receptor protein signaling pathway
protein transport
protein ubiquitination
regulation of cAMP biosynthetic process
regulation of cAMP catabolic process
negative regulation of protein ubiquitination
positive regulation of protein ubiquitination
receptor internalization
negative regulation of NF-kappaB transcription factor activity
positive regulation of synaptic transmission, dopaminergic
negative regulation of interleukin-1 beta production
negative regulation of interleukin-12 production
negative regulation of interleukin-6 production
negative regulation of tumor necrosis factor production
positive regulation of peptidyl-serine phosphorylation
negative regulation of toll-like receptor signaling pathway
negative regulation of GTPase activity
negative regulation of smooth muscle cell apoptotic process
follicle-stimulating hormone signaling pathway
negative regulation of cysteine-type endopeptidase activity involved in apoptotic process
proteasome-mediated ubiquitin-dependent protein catabolic process
negative regulation of natural killer cell mediated cytotoxicity
positive regulation of peptidyl-tyrosine phosphorylation
detection of temperature stimulus involved in sensory perception of pain
positive regulation of protein kinase B signaling
positive regulation of calcium ion transport
cell chemotaxis
regulation of androgen receptor signaling pathway
positive regulation of ERK1 and ERK2 cascade
negative regulation of release of cytochrome c from mitochondria
positive regulation of DNA biosynthetic process
G-protein coupled receptor binding
receptor binding
protein binding
enzyme binding
protein domain specific binding
clathrin binding
ubiquitin protein ligase binding
alpha-1A adrenergic receptor binding
alpha-1B adrenergic receptor binding
angiotensin receptor binding
type 1 angiotensin receptor binding
D1 dopamine receptor binding
follicle-stimulating hormone receptor binding
type 2A serotonin receptor binding
protein complex binding
protein complex scaffold
protein kinase B binding
mitogen-activated protein kinase binding
14-3-3 protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-vnb2r:80/100.66.79.197:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0