Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other BLNK SIRNA Assays ›
          Gene Symbol:
          BLNK
          Gene Name:
          B-cell linker
          Gene Aliases:
          AGM4, BASH, BLNK-S, LY57, SLP-65, SLP65, bca
          Chromosome Location:
          Chr.10: 96189171 - 96271576 on Build GRCh38
          UniGene ID:
          Hs.665244
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 20210

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: BLNK
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001114094.1 NP_001107566.1 3, 4 335
          NM_001258440.1 NP_001245369.1 3, 4 275
          NM_001258441.1 NP_001245370.1 3, 4 275
          NM_001258442.1 NP_001245371.1 3, 4 275
          NM_013314.3 NP_037446.1 3, 4 335
          NR_047680.1 3, 4 275
          NR_047682.1 3, 4 275
          NR_047683.1 3, 4 275
          XM_011539728.2 XP_011538030.1 3, 4 212
          XM_017016159.1 XP_016871648.1 3, 4 213
          GenBank mRNA AF068180.1 - 3, 4 310
          AF068181.1 - 3, 4 310
          AK027779.1 - 3, 4 270
          AK225546.1 - 3, 4 318
          AM180327.1 - 3, 4 157
          AM180328.1 - 3, 4 157
          AM180329.1 - 3, 4 157
          AM180330.1 - 3, 4 157
          AM180331.1 - 3, 4 157
          AM180332.1 - 3, 4 157
          AM180333.1 - 3, 4 157
          AM180334.1 - 3, 4 157
          AM180336.1 - 3, 4 157
          AM180337.1 - 3, 4 157
          AM180339.1 - 3, 4 157
          AM180340.1 - 3, 4 157
          BC018906.2 - 3, 4 249 4053894
          BC110298.1 - 3, 4 267
          BM820310.1 - 3, 4 328
          BU597768.1 - 3, 4 280
          DA045252.1 - 3, 4 335

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          scaffold/adaptor protein

          Gene Ontology Categories:

          Function(s) Process(es)

          inflammatory response
          humoral immune response
          transmembrane receptor protein tyrosine kinase signaling pathway
          positive regulation of signal transduction
          B cell differentiation
          intracellular signal transduction
          transmembrane receptor protein tyrosine kinase adaptor activity
          SH3/SH2 adaptor activity
          protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-5688f:80/100.66.73.65:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0