Search
Search

POC1B-GALNT4
GALNT4| Gene Symbol: POC1B-GALNT4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001199781.1 | NP_001186710.1 | 3 | 1634 | ||
| NM_001199782.1 | NP_001186711.1 | 3 | 867 | |||
| GenBank mRNA | AK297677.1 | - | 3 | 867 | ||
| BX647258.1 | - | 3 | 1370 | |||
| Gene Symbol: GALNT4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003774.4 | NP_003765.2 | 1 | 1533 | ||
| GenBank mRNA | AK312870.1 | - | 1 | 1483 | ||
| AW835269.1 | - | 1 | 100 | |||
| BC036390.2 | - | 1 | 1489 | 5261236 | ||