Search
Search

NARS| Gene Symbol: NARS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004539.3 | NP_004530.1 | 2 | 298 | ||
| XM_005266700.1 | XP_005266757.1 | 2 | 76 | |||
| GenBank mRNA | AJ000334.1 | - | 2 | 143 | ||
| AK129897.1 | - | 2 | 101 | |||
| AK222835.1 | - | 2 | 145 | |||
| AK294364.1 | - | 2 | 145 | |||
| AK294763.1 | - | 2 | 129 | |||
| AK297777.1 | - | 2 | 138 | |||
| BC001687.1 | - | 2 | 147 | 2987840 | ||
| D84273.1 | - | 2 | 102 | |||
| DC302277.1 | - | 2 | 299 | |||