Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other INS1 SIRNA Assays ›
Gene Symbol:
Ins1
Gene Name:
insulin 1
Gene Aliases:
-
Chromosome Location:
Chr.1: 272799784 - 272800351 on Build Rnor_6.0
UniGene ID:
Rn.962
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
216170
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Ins1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_019129.3
NP_062002.1
2
328
GenBank mRNA
BP465191.1
-
1
280
C07095.1
-
1
127
CB702626.1
-
2
315
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_008386, AK148541, BC145868, AK007345, AK007482, and others.
Panther Classification:
Molecular Function -
growth factor
Gene Ontology Categories:
Function(s)
Process(es)
MAPK cascade
negative regulation of acute inflammatory response
glucose metabolic process
regulation of cellular amino acid metabolic process
acute-phase response
G-protein coupled receptor signaling pathway
positive regulation of cell proliferation
insulin receptor signaling pathway
response to organic substance
positive regulation of phosphatidylinositol 3-kinase signaling
glucose transport
regulation of transmembrane transporter activity
activation of protein kinase B activity
positive regulation of cellular protein metabolic process
negative regulation of protein oligomerization
regulation of protein localization
negative regulation of NAD(P)H oxidase activity
response to cytokine
wound healing
negative regulation of protein catabolic process
glucose homeostasis
positive regulation of MAPK cascade
response to peptide hormone
positive regulation of glycogen biosynthetic process
positive regulation of DNA replication
negative regulation of glycogen catabolic process
positive regulation of glycolytic process
positive regulation of mitotic nuclear division
negative regulation of proteolysis
negative regulation of fatty acid metabolic process
positive regulation of glucose import
positive regulation of insulin receptor signaling pathway
alpha-beta T cell activation
regulation of protein secretion
negative regulation of protein secretion
positive regulation of cytokine secretion
positive regulation of peptidyl-tyrosine phosphorylation
negative regulation of lipid catabolic process
positive regulation of NF-kappaB transcription factor activity
response to cAMP
positive regulation of protein kinase B signaling
fatty acid homeostasis
negative regulation of respiratory burst involved in inflammatory response
positive regulation of respiratory burst
positive regulation of protein localization to nucleus
negative regulation of feeding behavior
protease binding
insulin receptor binding
insulin-like growth factor receptor binding
hormone activity
identical protein binding
chaperone binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0