Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MED24 SIRNA Assays ›
Gene Symbol:
MED24
Gene Name:
mediator complex subunit 24
Gene Aliases:
ARC100, CRSP100, CRSP4, DRIP100, MED5, THRAP4, TRAP100
Chromosome Location:
Chr.17: 40019097 - 40054636 on Build GRCh38
UniGene ID:
Hs.462983
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
22155
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
4,126.00
Your Price
Online offer:
4126.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: MED24
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001079518.1
NP_001072986.1
4
589
NM_001267797.1
NP_001254726.1
4
567
NM_014815.3
NP_055630.2
5
606
XM_005257874.2
XP_005257931.1
5
511
XM_006722204.2
XP_006722267.1
5
511
XM_006722206.2
XP_006722269.1
5
419
XM_006722207.2
XP_006722270.1
5
387
XM_011525529.2
XP_011523831.1
5
511
XM_011525530.2
XP_011523832.1
5
387
XM_011525531.2
XP_011523833.1
5
387
XM_011525532.2
XP_011523834.1
5
419
XM_011525533.2
XP_011523835.1
5
387
XM_011525534.2
XP_011523836.1
4
330
XM_011525535.2
XP_011523837.1
4
330
XM_011525536.2
XP_011523838.1
5
387
XM_011525538.2
XP_011523840.1
4
330
XM_017025458.1
XP_016880947.1
5
419
XM_017025459.1
XP_016880948.1
5
419
XM_017025460.1
XP_016880949.1
5
419
XM_017025461.1
XP_016880950.1
5
511
XM_017025462.1
XP_016880951.1
5
419
XM_017025463.1
XP_016880952.1
5
387
XM_017025464.1
XP_016880953.1
5
387
XM_017025465.1
XP_016880954.1
5
511
XM_017025466.1
XP_016880955.1
5
419
XM_017025467.1
XP_016880956.1
5
419
XM_017025468.1
XP_016880957.1
5
387
XM_017025469.1
XP_016880958.1
4
330
XM_017025470.1
XP_016880959.1
4
330
XM_017025471.1
XP_016880960.1
4
330
GenBank mRNA
AF055995.1
-
4
293
AF277379.1
-
4
293
AK022508.1
-
4
358
AK291040.1
-
4
338
AK293309.1
-
3
128
AK300992.1
-
4
211
BC011375.1
-
4
568
3866038
D50920.1
-
5
366
EU392534.1
-
3
160
EU392535.1
-
1
56
EU392536.1
-
4
548
EU392539.1
-
3
160
EU392540.1
-
3
160
EU392543.1
-
3
160
EU392545.1
-
3
160
EU392548.1
-
1
56
EU392552.1
-
4
310
EU392553.1
-
3
160
Back To Top
More Information
Panther Classification:
Molecular Function -
general transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
regulation of transcription from RNA polymerase II promoter
transcription initiation from RNA polymerase II promoter
protein ubiquitination
histone acetylation
stem cell population maintenance
intracellular steroid hormone receptor signaling pathway
androgen receptor signaling pathway
positive regulation of transcription, DNA-templated
protein heterooligomerization
RNA polymerase II transcription cofactor activity
transcription cofactor activity
histone acetyltransferase activity
receptor activity
protein binding
ligand-dependent nuclear receptor transcription coactivator activity
vitamin D receptor binding
thyroid hormone receptor binding
ubiquitin protein ligase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0