Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNF44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014901.4 | NP_055716.1 | 11 | 2608 | ||
XM_005265840.2 | XP_005265897.1 | 11 | 2320 | |||
XM_005265841.4 | XP_005265898.1 | 11 | 2565 | |||
XM_005265842.4 | XP_005265899.1 | 11 | 2550 | |||
XM_005265843.2 | XP_005265900.1 | 11 | 2256 | |||
XM_005265844.3 | XP_005265901.1 | 11 | 2361 | |||
XM_005265845.4 | XP_005265902.1 | 11 | 2499 | |||
XM_006714831.2 | XP_006714894.1 | 11 | 2311 | |||
XM_006714832.2 | XP_006714895.1 | 11 | 2179 | |||
XM_011534466.2 | XP_011532768.1 | 11 | 2323 | |||
XM_011534467.2 | XP_011532769.1 | 11 | 2554 | |||
GenBank mRNA | AB029023.1 | - | 11 | 2493 | ||
AK127673.1 | - | 2 | 1430 | |||
BC039833.1 | - | 11 | 2564 | 5535911 | ||
BC063297.1 | - | 11 | 2609 | 30349418 |