Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CDKN1B SIRNA Assays ›
Gene Symbol:
CDKN1B
Gene Name:
cyclin dependent kinase inhibitor 1B
Gene Aliases:
CDKN4, KIP1, MEN1B, MEN4, P27KIP1
Chromosome Location:
Chr.12: 12717270 - 12722383 on Build GRCh38
UniGene ID:
Hs.238990
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
242378
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CDKN1B
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_004064.4
NP_004055.1
3
2138
GenBank mRNA
AA833962.1
-
1
162
AY004255.1
-
3
1720
BC001971.1
-
3
1945
3458141
Back To Top
More Information
Panther Classification:
Molecular Function -
kinase inhibitor
Gene Ontology Categories:
Function(s)
Process(es)
regulation of cyclin-dependent protein serine/threonine kinase activity
G1/S transition of mitotic cell cycle
response to hypoxia
potassium ion transport
activation of cysteine-type endopeptidase activity involved in apoptotic process
DNA damage response, signal transduction by p53 class mediator resulting in cell cycle arrest
cell cycle arrest
Notch signaling pathway
sensory perception of sound
positive regulation of cell proliferation
negative regulation of cell proliferation
response to glucose
positive regulation of cell death
negative regulation of cell growth
positive regulation of microtubule polymerization
response to estradiol
negative regulation of kinase activity
negative regulation of phosphorylation
response to drug
negative regulation of apoptotic process
response to amino acid
response to peptide hormone
positive regulation of protein catabolic process
negative regulation of cyclin-dependent protein serine/threonine kinase activity
positive regulation of cyclin-dependent protein serine/threonine kinase activity
positive regulation of cell cycle
negative regulation of transcription, DNA-templated
negative regulation of mitotic cell cycle
response to cadmium ion
autophagic cell death
inner ear development
negative regulation of cellular component movement
negative regulation of epithelial cell proliferation involved in prostate gland development
cellular response to antibiotic
cellular response to lithium ion
cellular response to organic cyclic compound
mitotic cell cycle arrest
negative regulation of vascular smooth muscle cell proliferation
cyclin-dependent protein serine/threonine kinase inhibitor activity
transforming growth factor beta receptor, cytoplasmic mediator activity
protein binding
cysteine-type endopeptidase activator activity involved in apoptotic process
protein phosphatase binding
Hsp70 protein binding
protein complex binding
chaperone binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0