Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CYP1B1 SIRNA Assays ›
Gene Symbol:
CYP1B1
Gene Name:
cytochrome P450 family 1 subfamily B member 1
Gene Aliases:
CP1B, CYPIB1, GLC3A, P4501B1
Chromosome Location:
Chr.2: 38067603 - 38076181 on Build GRCh38
UniGene ID:
Hs.154654
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
2439
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CYP1B1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_000104.3
NP_000095.2
3
2270
GenBank mRNA
BC012049.1
-
3
2223
4662252
U03688.1
-
3
2213
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
angiogenesis
trabecular meshwork development
cellular aromatic compound metabolic process
xenobiotic metabolic process
nitric oxide biosynthetic process
cell adhesion
visual perception
steroid metabolic process
estrogen metabolic process
negative regulation of cell proliferation
intrinsic apoptotic signaling pathway in response to oxidative stress
toxin metabolic process
response to toxic substance
positive regulation of vascular endothelial growth factor production
sterol metabolic process
arachidonic acid metabolic process
epoxygenase P450 pathway
collagen fibril organization
negative regulation of cell migration
negative regulation of NF-kappaB transcription factor activity
negative regulation of cell adhesion mediated by integrin
retinol metabolic process
retinal metabolic process
positive regulation of apoptotic process
endothelial cell migration
positive regulation of angiogenesis
positive regulation of JAK-STAT cascade
membrane lipid catabolic process
blood vessel morphogenesis
oxidation-reduction process
retinal blood vessel morphogenesis
cellular response to hydrogen peroxide
cellular response to organic cyclic compound
endothelial cell-cell adhesion
omega-hydroxylase P450 pathway
regulation of reactive oxygen species metabolic process
monooxygenase activity
iron ion binding
oxidoreductase activity, acting on paired donors, with incorporation or reduction of molecular oxygen, reduced flavin or flavoprotein as one donor, and incorporation of one atom of oxygen
oxygen binding
heme binding
aromatase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0