Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GUSBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027028.2 | 4 | 330 | |||
GenBank mRNA | AK290693.1 | - | 4 | 297 | ||
BC147000.1 | - | 4 | 284 |
Gene Symbol: GUSBP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
GenBank mRNA | AK289965.1 | - | 3 | 421 | ||
U21555.1 | - | LIT2100 5G4 pseudogene |
4 | 360 |
Gene Symbol: SMA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC131740.1 | - | 2 | 181 |