Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RASL12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001307930.1 | NP_001294859.1 | 4 | 1011 | ||
NM_016563.3 | NP_057647.1 | 5 | 1068 | |||
XM_011521660.2 | XP_011519962.1 | 6 | 881 | |||
GenBank mRNA | AF233588.1 | - | 5 | 907 | ||
AK296261.1 | - | 4 | 821 | |||
AK300536.1 | - | 5 | 851 | |||
AK314914.1 | - | 5 | 876 | |||
BC053734.1 | - | 5 | 916 | 5588236 |