Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other NTRK3 SIRNA Assays ›
          Gene Symbol:
          NTRK3
          Gene Name:
          neurotrophic receptor tyrosine kinase 3
          Gene Aliases:
          GP145-TrkC, TRKC, gp145(trkC)
          Chromosome Location:
          Chr.15: 87859749 - 88256796 on Build GRCh38
          UniGene ID:
          Hs.185701
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 248

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: NTRK3
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001007156.2 NP_001007157.1 3, 4 547
          NM_001012338.2 NP_001012338.1 3, 4 547
          NM_001243101.1 NP_001230030.1 3, 4 547
          NM_001320134.1 NP_001307063.1 1, 2 257
          NM_002530.3 NP_002521.2 3, 4 547
          XM_006720543.3 XP_006720606.1 1, 2 856
          XM_006720544.3 XP_006720607.1 1, 2 856
          XM_006720545.3 XP_006720608.1 1, 2 856
          XM_006720548.3 XP_006720611.1 1, 2 856
          XM_006720549.3 XP_006720612.1 1, 2 856
          XM_006720550.3 XP_006720613.1 1, 2 856
          XM_011521637.2 XP_011519939.1 1, 2 856
          XM_011521638.2 XP_011519940.1 1, 2 856
          XM_017022240.1 XP_016877729.1 2, 3 727
          XM_017022242.1 XP_016877731.1 1, 2 856
          XM_017022247.1 XP_016877736.1 1, 2 856
          XM_017022248.1 XP_016877737.1 1, 2 856
          XM_017022249.1 XP_016877738.1 1, 2 856
          XM_017022250.1 XP_016877739.1 1, 2 856
          XM_017022253.1 XP_016877742.1 1, 2 856
          XM_017022254.1 XP_016877743.1 1, 2 856
          GenBank mRNA AK297160.1 - 1, 2 265
          AK309695.1 - 1, 2 267
          AK313646.1 - 1, 2 266
          BC013693.1 - 1, 2 246 4181574
          BI962878.1 - 517
          BT007291.1 - 1, 2 241
          DA789502.1 - 3, 4 547
          KJ534904.1 - 1, 2 241
          KJ534905.1 - 1, 2 241
          KJ534906.1 - 1, 2 241
          KJ535052.1 - 1, 2 241
          S76475.1 - 1, 2 256
          S76476.1 - 1, 2 256
          U05012.1 - 2, 3 396

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_001270655, NM_019248, NM_001270656, XM_008759513, L14445, and others.
          This siRNA also targets the following transcripts in Mouse: NM_008746, NM_182809, JF417977, AF035399, AY336094, and others.

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          activation of MAPK activity
          neuron migration
          negative regulation of protein phosphorylation
          positive regulation of protein phosphorylation
          transmembrane receptor protein tyrosine kinase signaling pathway
          heart development
          circadian rhythm
          positive regulation of cell proliferation
          positive regulation of gene expression
          peptidyl-tyrosine phosphorylation
          modulation by virus of host transcription
          positive regulation of cell migration
          activation of protein kinase B activity
          positive regulation of peptidyl-serine phosphorylation
          neurotrophin signaling pathway
          mechanoreceptor differentiation
          positive regulation of apoptotic process
          response to ethanol
          protein autophosphorylation
          ephrin receptor signaling pathway
          neuron fate specification
          response to axon injury
          positive regulation of axon extension involved in regeneration
          negative regulation of astrocyte differentiation
          positive regulation of positive chemotaxis
          response to corticosterone
          positive regulation of synapse assembly
          negative regulation of cell death
          lens fiber cell differentiation
          cellular response to retinoic acid
          cochlea development
          activation of GTPase activity
          positive regulation of actin cytoskeleton reorganization
          p53 binding
          GPI-linked ephrin receptor activity
          neurotrophin receptor activity
          protein binding
          ATP binding
          neurotrophin binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0