Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcnq5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001160139.1 | NP_001153611.1 | 3 | 894 | ||
NM_023872.3 | NP_076361.1 | 3 | 894 | |||
GenBank mRNA | AF263836.1 | - | 3 | 369 | ||
AK147264.1 | - | 3 | 897 | |||
AY679158.1 | - | 3 | 533 |