Search
Search

Ndufs6| Gene Symbol: Ndufs6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010888.2 | NP_035018.1 | 4 | 421 | ||
| GenBank mRNA | AK147147.1 | - | 4 | 451 | ||
| BC059730.1 | - | 4 | 394 | |||
| BC076501.1 | - | 4 | 452 | |||
| BC086933.1 | - | 4 | 394 | 6760537 | ||
| BG081040.2 | - | 4 | 411 | |||