Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NAV2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001244963.1 | NP_001231892.1 | 1 | 507 | ||
NM_145117.4 | NP_660093.2 | 1 | 507 | |||
NM_182964.5 | NP_892009.3 | 1 | 507 | |||
XM_006718364.3 | XP_006718427.1 | 1 | 829 | |||
XM_006718365.3 | XP_006718428.1 | 1 | 829 | |||
XM_011520444.2 | XP_011518746.1 | 1 | 829 | |||
XM_011520445.2 | XP_011518747.1 | 1 | 829 | |||
XM_011520446.2 | XP_011518748.1 | 1 | 829 | |||
XM_011520447.2 | XP_011518749.1 | 1 | 829 | |||
XM_011520449.2 | XP_011518751.1 | 1 | 829 | |||
XM_011520450.2 | XP_011518752.1 | 1 | 829 | |||
XM_017018521.1 | XP_016874010.1 | 1 | 829 | |||
GenBank mRNA | AB037840.3 | - | 1 | 508 | ||
AB063116.1 | - | 1 | 476 | |||
AF466143.1 | - | 1 | 222 | |||
AF466144.1 | - | 1 | 222 | |||
AJ488102.1 | - | 1 | 245 | |||
BC150290.1 | - | 1 | 508 | 8860130 |