Search
Search

PVR| Gene Symbol: PVR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135768.2 | NP_001129240.1 | 8 | 5452 | ||
| NM_001135769.2 | NP_001129241.1 | 7 | 5428 | |||
| NM_006505.4 | NP_006496.4 | 8 | 5587 | |||
| GenBank mRNA | AK098812.1 | - | 1 | 1714 | ||