Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other VANGL2 SIRNA Assays ›
          Gene Symbol:
          VANGL2
          Gene Name:
          VANGL planar cell polarity protein 2
          Gene Aliases:
          LPP1, LTAP, STB1, STBM, STBM1
          Chromosome Location:
          Chr.1: 160400574 - 160428678 on Build GRCh38
          UniGene ID:
          Hs.99477
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 279802

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: VANGL2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_020335.2 NP_065068.1 8 2759
          XM_005245357.1 XP_005245414.1 8 2566
          XM_011509804.1 XP_011508106.1 8 2614
          GenBank mRNA AB033041.1 - 7 2435

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          establishment of planar polarity
          neural tube closure
          hair follicle development
          heart looping
          membranous septum morphogenesis
          muscular septum morphogenesis
          planar cell polarity pathway involved in axis elongation
          Rho protein signal transduction
          anterior/posterior pattern specification
          heparan sulfate proteoglycan biosynthetic process
          convergent extension involved in neural plate elongation
          regulation of Wnt signaling pathway
          glomerulus development
          regulation of actin cytoskeleton organization
          somatic stem cell population maintenance
          nonmotile primary cilium assembly
          cell migration involved in kidney development
          post-anal tail morphogenesis
          dopaminergic neuron axon guidance
          serotonergic neuron axon guidance
          wound healing
          positive regulation of JUN kinase activity
          apical protein localization
          establishment or maintenance of epithelial cell apical/basal polarity
          somatic stem cell division
          establishment of body hair planar orientation
          digestive tract morphogenesis
          convergent extension involved in axis elongation
          inner ear receptor stereocilium organization
          orthogonal dichotomous subdivision of terminal units involved in lung branching morphogenesis
          planar dichotomous subdivision of terminal units involved in lung branching morphogenesis
          lateral sprouting involved in lung morphogenesis
          kidney morphogenesis
          planar cell polarity pathway involved in heart morphogenesis
          cochlea morphogenesis
          establishment of planar polarity involved in neural tube closure
          planar cell polarity pathway involved in neural tube closure
          protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-vnb2r:80/100.66.79.197:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0