Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: CASS4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164114.1 | NP_001157586.1 | 5 | 732 | ||
NM_001164116.1 | NP_001157588.1 | 6 | 918 | |||
NM_020356.3 | NP_065089.2 | 5 | 894 | |||
XM_006723831.3 | XP_006723894.1 | 6 | 1318 | |||
GenBank mRNA | AJ276678.1 | - | 6 | 918 | ||
AK295613.1 | - | 5 | 597 | |||
AK308057.1 | - | 5 | 895 | |||
AK314653.1 | - | 5 | 827 | |||
BC027951.1 | - | 5 | 844 | 5205865 |