Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Elmod1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177769.4 | NP_808437.2 | 4 | 537 | ||
GenBank mRNA | AK029207.1 | - | 4 | 419 | ||
AK132239.1 | - | 4 | 538 | |||
BC049160.1 | - | 4 | 400 | |||
BC120566.1 | - | 3 | 253 | |||
BC144961.1 | - | 3 | 253 | |||
BC144963.1 | - | 3 | 215 | 9053511 |