Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RRAGD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021244.4 | NP_067067.1 | 1, 2 | 435 | ||
XM_005248755.4 | XP_005248812.1 | 1, 2 | 629 | |||
GenBank mRNA | AF272036.1 | - | 1, 2 | 218 | ||
AK289799.1 | - | 1, 2 | 360 | |||
AL523999.3 | - | 1, 2 | 435 | |||
BC003088.1 | - | 1, 2 | 97 | 3536088 | ||
DB472739.1 | - | 1, 2 | 427 |