Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: ANP32B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006401.2 | NP_006392.1 | 3 | 592 | ||
GenBank mRNA | AK223454.1 | - | 3 | 459 | ||
AK313733.1 | - | 3 | 447 | |||
BC013003.2 | - | 3 | 424 | 3010189 | ||
BC019658.1 | - | 3 | 431 | 4478860 | ||
U70439.1 | - | 3 | 453 | |||
Y07569.1 | - | 3 | 298 | |||
Y07570.1 | - | 3 | 427 | |||
Y07969.1 | - | 3 | 467 |