Search
Search

WDR24| Gene Symbol: WDR24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032259.3 | NP_115635.1 | 1 | 960 | ||
| XM_011522699.2 | XP_011521001.1 | 1 | 1197 | |||
| GenBank mRNA | AK301146.1 | - | 1 | 944 | ||
| AL136863.1 | - | 1 | 937 | |||
| BC008025.1 | - | 1 | 425 | 3544791 | ||