Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other BTRC SIRNA Assays ›
          Gene Symbol:
          BTRC
          Gene Name:
          beta-transducin repeat containing E3 ubiquitin protein ligase
          Gene Aliases:
          BETA-TRCP, FBW1A, FBXW1, FBXW1A, FWD1, bTrCP, bTrCP1, betaTrCP
          Chromosome Location:
          Chr.10: 101353808 - 101557321 on Build GRCh38
          UniGene ID:
          Hs.643802
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 35486

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: BTRC
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001256856.1 NP_001243785.1 4 480
          NM_003939.4 NP_003930.1 4 450
          NM_033637.3 NP_378663.1 5 558
          XM_006718054.2 XP_006718117.1 5 858
          XM_011540320.2 XP_011538622.1 2 294
          XM_017016870.1 XP_016872359.1 4 780
          XM_017016871.1 XP_016872360.1 5 449
          XM_017016872.1 XP_016872361.1 5 621
          XM_017016873.1 XP_016872362.1 5 477
          XM_017016874.1 XP_016872363.1 3 372
          GenBank mRNA AF101784.1 - 5 410
          AF129530.1 - 4 371
          AK295870.1 - 4 348
          AK313353.1 - 5 431
          AK313417.1 - 4 414
          BC027994.1 - 5 447 5180993
          CR749295.1 - 4 411
          GU727631.1 - 5 447
          KU178451.1 - 5 410
          KU178452.1 - 7 559
          KU178453.1 - 6 503
          KU178454.1 - 4 332
          Y14153.1 - 4 371

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          G2/M transition of mitotic cell cycle
          protein polyubiquitination
          stimulatory C-type lectin receptor signaling pathway
          protein dephosphorylation
          ubiquitin-dependent protein catabolic process
          signal transduction
          viral process
          Wnt signaling pathway
          protein ubiquitination
          SCF-dependent proteasomal ubiquitin-dependent protein catabolic process
          protein destabilization
          mammary gland epithelial cell proliferation
          NIK/NF-kappaB signaling
          Fc-epsilon receptor signaling pathway
          regulation of circadian rhythm
          positive regulation of circadian rhythm
          regulation of I-kappaB kinase/NF-kappaB signaling
          proteasome-mediated ubiquitin-dependent protein catabolic process
          negative regulation of sequence-specific DNA binding transcription factor activity
          positive regulation of proteolysis
          negative regulation of smoothened signaling pathway
          negative regulation of transcription, DNA-templated
          positive regulation of transcription, DNA-templated
          rhythmic process
          T cell receptor signaling pathway
          stress-activated MAPK cascade
          positive regulation of ubiquitin-protein ligase activity involved in regulation of mitotic cell cycle transition
          branching involved in mammary gland duct morphogenesis
          regulation of canonical Wnt signaling pathway
          regulation of proteasomal protein catabolic process
          cellular response to organic cyclic compound
          ubiquitin-protein transferase activity
          protein binding
          beta-catenin binding
          ligase activity
          protein phosphorylated amino acid binding
          protein dimerization activity
          ubiquitin protein ligase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-st8h5:80/100.66.79.144:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0