Search
Search

ABCA9| Gene Symbol: ABCA9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080283.3 | NP_525022.2 | 10 | 1494 | ||
| XM_017024011.1 | XP_016879500.1 | 10 | 1459 | |||
| XM_017024012.1 | XP_016879501.1 | 10 | 1563 | |||
| XM_017024013.1 | XP_016879502.1 | 10 | 1383 | |||
| XM_017024014.1 | XP_016879503.1 | 10 | 1983 | |||
| XM_017024015.1 | XP_016879504.1 | 7 | 1006 | |||
| GenBank mRNA | AF423307.1 | - | 9 | 1351 | ||
| AK057068.1 | - | 10 | 1426 | |||
| AK074491.1 | - | 3 | 303 | |||
| AY028899.1 | - | 10 | 1494 | |||
| CR627382.1 | - | 9 | 1300 | |||